Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-tert-butyl Disulfide 97.0+%, TCI America™
SDP

Catalog No. D0225100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0225100ML 100 mL
D022525ML 25 mL
D0225500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0225100ML Supplier TCI America Supplier No. D0225100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-06-5
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00008838
InChI Key BKCNDTDWDGQHSD-UHFFFAOYSA-N
Synonym di-tert-butyl disulfide, disulfide, bis 1,1-dimethylethyl, 1,2-di-tert-butyldisulfane, di-tert-butyl disulphide, tert-butyl disulfide, t-butyl disulfide, 2-tert-butyldisulfanyl-2-methylpropane, di-t-butyl disulfide, t-butyldisulfide, 2,2'-dithiobis 2-methylpropane
PubChem CID 66060
IUPAC Name 2-(tert-butyldisulfanyl)-2-methylpropane
SMILES CC(C)(C)SSC(C)(C)C

Specifications

Color Yellow
Boiling Point 200°C
Quantity 100 mL
UN Number 3082
Formula Weight 178.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-tert-butyl Disulfide
EINECSNumber (2)-0477
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.