Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetoxydimethylsilane, 95%, Thermo Scientific™

Catalog No. AC426690250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426690250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426690250 Supplier Thermo Scientific Chemicals Supplier No. 426690250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 2182-66-3
Molecular Formula C6H12O4Si
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039832
InChI Key RQVFGTYFBUVGOP-UHFFFAOYSA-N
Synonym dimethyldiacetoxysilane, diacetoxydimethylsilane, dimethylsilanediyl diacetate, silanediol, dimethyl-, diacetate, dimethylsilanediol, diacetate, silanediol, 1,1-dimethyl-, 1,1-diacetate, acetyloxy dimethylsilyl acetate, dimethyldiacetoxysilicane, dimethyl-silanediodiacetate, acmc-1ak12
PubChem CID 75130
IUPAC Name [acetyloxy(dimethyl)silyl] acetate
SMILES CC(=O)O[Si](C)(C)OC(=O)C

Specifications

Melting Point -12.5°C
Color Colorless
Density 1.0530g/mL
Boiling Point 164°C
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
Linear Formula (CH3COO)2Si(CH3)2
Quantity 25 g
Specific Gravity 1.053
Formula Weight 176.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diacetoxydimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
EINECSNumber 218-562-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.