Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diacetyl-D-tartaric Anhydride 98.0+%, TCI America™
SDP

Catalog No. D264525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D264525G 25 g
D26455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D264525G Supplier TCI America Supplier No. D264525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 70728-23-3
Molecular Formula C8H8O7
Molecular Weight (g/mol) 216.145
MDL Number MFCD02093461
InChI Key XAKITKDHDMPGPW-WDSKDSINSA-N
PubChem CID 12731855
IUPAC Name [(3S,4S)-4-acetyloxy-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1C(C(=O)OC1=O)OC(=O)C

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 216.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Diacetyl-D-tartaric Anhydride
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.