Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetyl Monoxime 98.0+%, TCI America™
SDP

Catalog No. B0683500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0683500G 500 g
B068325G 25 g
B0683100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0683500G Supplier TCI America Supplier No. B0683500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-(hydroxyimino)butan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 186°C
Quantity 500 g
Formula Weight 101.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diacetyl Monoxime
EINECSNumber (9)-0538
RTECSNumber EK3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.