Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. D3768250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3768250G 250 g
D376825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3768250G Supplier TCI America Supplier No. D3768250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20306-22-3
Molecular Formula C14H20O4
Molecular Weight (g/mol) 252.31
MDL Number MFCD00050452
InChI Key ZXTPJLLIHIDBKQ-UHFFFAOYSA-N
Synonym 1,4-Cyclohexanedicarboxylic Acid Diallyl Ester
PubChem CID 11265237
IUPAC Name bis(prop-2-enyl) cyclohexane-1,4-dicarboxylate
SMILES C=CCOC(=O)C1CCC(CC1)C(=O)OCC=C

Specifications

Color Yellow
Quantity 250 g
Formula Weight 252.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl 1,4-Cyclohexanedicarboxylate (cis- and trans- mixture)
EINECSNumber (3)-4376
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.