Learn More
Lkt Laboratories CAS 2444-49-7, ≥90%, Organosulfur found in garlic; Microtubule polymerization inhibitor.
SDP
Supplier:  Lkt Laboratories D3203
Encompass_Preferred
Diallyl tetrasulfide is an organosulfur that displays anticancer, antioxidative, and antimicrobial properties. In vitro, diallyl tetrasulfide increases reactive oxygen species (ROS) and heme oxygenase-1 levels and inhibits microtubule polymerization, inducing mitotic arrest. Diallyl tetrasulfide exhibits antibacterial and antifungal benefit against many microbes, including including Staphylococcus aureus, methicillin-resistant Staphylococcus aureus (MRSA), and several species of Candida and Aspergillus. Additionally, this compound decreases cadmium-mediated oxidative damage in several animal models, increasing levels of superoxide dismutase, glutathione peroxidase, and other glutathione metabolizing enzymes.
Catalog No. NC2535082
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.