Learn More
Abcam Diallyl Tetrasulfide, ROS Inducer, >90% Purity, Liquid, 5 mg
Description
- Molecular Weight: 210.4 Da
- Purity: >90%
- Biological Activity: JNK activator, ROS inducer, irreversible Cdc25 isoforms A and C inhibitor, and tubulin target
- Biological Effects: Induces G2-M arrest and apoptosis; displays antiproliferative, cytoprotective, and antioxidant effects in vivo
- Form: Liquid
- Constituents: 100% diallyltetrasulfane
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Tubulin, Cdc25 |
| Form | Liquid |
| Shipping Condition | Blue Ice |
| Product Type | Antioxidant |
| Buffer | 100% diallyltetrasulfane |
| Content And Storage | Store at -20°C. |
| Formulation | 100% diallyltetrasulfane |
| Quantity | 5 mg |
| For Use With (Application) | JNK activation, ROS induction, Cdc25 inhibitor, apoptosis induction, G2-M arrest |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.