Learn More
Abcam Diallyl Tetrasulfide, ROS Inducer, Liquid, >90% Purity, 100 mg
Description
- Molecular Weight: 210.4 Da
- Purity: >90%
- Form: Liquid
- Key Biological Effects: ROS inducer, JNK activator, irreversible Cdc25A and Cdc25C inhibitor, and targets tubulin
- Functional Outcomes: Induces G2-M arrest and apoptosis; displays antiproliferative, cytoprotective, and antioxidant effects in vivo
- Other: Orally active
- Composition: 100% diallyltetrasulfane
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Liquid |
| Content And Storage | Store at -20°C. |
| Product Type | Antioxidant |
| Buffer | 100% diallyltetrasulfane |
| Shipping Condition | Blue Ice |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Tubulin |
| Formulation | 100% diallyltetrasulfane |
| Quantity | 100 mg |
| For Use With (Application) | JNK activation, ROS induction, Cdc25 inhibition, G2-M arrest, apoptosis induction, antiproliferative research, cytoprotective research, antioxidant research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.