Learn More
Medchemexpress LLC HY-117235 50mg Medchemexpress, Diallyl Trisulfide CAS:2050-87-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11723550MG
Encompass_Preferred
Medchemexpress, HY-117235 50mg Diallyl Trisulfide CAS:2050-87-5 Diallyl Trisulfide is isolated from Garlic. Diallyl Trisulfide suppresses the growth of Penicillium expansum (MFC99 value: ≤ 90 μg/mL) and promotes apoptosis via production of reactive oxygen species (ROS) and disintegration of cellular ultrastructure. Anticancer effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0900
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.