Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallylmethylamine, 98%

Catalog No. AC456340050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456340050 5 g
AC456340250 25 g
AC456341000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC456340050 Supplier Thermo Scientific Chemicals Supplier No. 456340050
Add to Cart
This item is not returnable. View return policy

CAS: 2424-01-3 | C7H13N | 111.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2424-01-3
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.18

Specifications

Density 0.7890g/mL
Boiling Point 111.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 0.789
Synonym 1,4-bis glycidyloxy benzene, hydroquinone diglycidyl ether, 2-4-oxiran-2-ylmethoxy phenoxymethyl oxirane, hydroquinone 1,4-diglycidyl ether, oxirane, 2,2'-1,4-phenylenebis oxymethylene bis, 2,2'-1,4-phenylenebis oxymethylene dioxirane, 2-4-oxiran-2-ylmethoxy phenoxy methyl oxirane, 1,4-bis oxiran-2-ylmethoxy benzene, acmc-20ap2z
InChI Key FSYPIGPPWAJCJG-UHFFFAOYNA-N
SMILES C1C(O1)COC2=CC=C(C=C2)OCC3CO3
IUPAC Name 2-[[4-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane
PubChem CID 62443
Formula Weight 111.18
Percent Purity 98%
Chemical Name or Material Diallylmethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 219-354-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.