Learn More
Diallylmethylamine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.7890g/mL |
| Boiling Point | 111.0°C |
| Flash Point | 7°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Quantity | 25 g |
| Specific Gravity | 0.789 |
| Synonym | diallylmethylamine, methyldiallylamine, 2-propen-1-amine, n-methyl-n-2-propenyl, n-allyl-n-methylprop-2-en-1-amine, methylbis prop-2-en-1-yl amine, methyldiprop-2-enylamine, 2-propen-1-amine, n-methyl-n-2-propen-1-yl, diallyl methylamine, n-methyldiallylamin, n-methyldiallylamine |
| InChI Key | WGESLFUSXZBFQF-UHFFFAOYSA-N |
| SMILES | CN(CC=C)CC=C |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call |
| EINECSNumber | 219-354-4 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.