Learn More
Biodesign Inc of Ny Dialysis Tubing, 14000 molecular weight cut-off, 6.4 mm wet diameter, 100 foot roll
Supplier:  Biodesign Inc of Ny D001
Encompass
Dialysis Tubing, 14000 molecular weight cut-off, 6.4 mm wet diameter, 100 foot roll
Catalog No. NC2545577
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.