Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamantane 98.0+%, TCI America™
SDP

Catalog No. D2876100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2876100MG 100 mg
D28761G 1 g
D28765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2876100MG Supplier TCI America Supplier No. D2876100MG

Chemical Identifiers

CAS 2292-79-7
Molecular Formula C14H20
Molecular Weight (g/mol) 188.314
MDL Number MFCD00171301
InChI Key ZICQBHNGXDOVJF-UHFFFAOYSA-N
Synonym Congressane, Pentacyclo[7.3.1.1(4,12).0(2,7).0(6,11)]tetradecane
PubChem CID 123154
ChEBI CHEBI:38223
SMILES C1C2CC3C4C1C5CC(C4)CC3C5C2

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 100 mg
Formula Weight 188.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.