Learn More
Diamedix™ 96-Well Assay Plates

Description
- Blank well plates for use with Diamedix test kits
- Each plate contains 96 wells
- Fisher Cat No. 23-451-055 is meant to be used with Diamedix Cardiolipins, Diamedix β2-Glycoproteins and tissue Transglutaminase test kits
- Fisher Cat No. 23-451-056 is meant to be used with Diamedix C6 Lyme test kit
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Black |
| No. of Wells | 96 |
| Surface Treatment | Non-treated |
| For Use With (Application) | Diamedix Kits (other than Cardiolipin, B2-Glycoprotein, Tissue Transglutaminase, C6 Lyme) |
| Quantity | 2/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.