Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Diamond RV-Pette PRO™ Sterile Dispenser Syringe Tips

Catalog No. 220101181
Change view
Click to view available options
Volume (Metric):
0.05 mL
0.1 mL
0.2 mL
0.5 mL
1.0 mL
1.25 mL
2.5 mL
5 mL
10 mL
12.5 mL
25 mL
50 mL
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-010-1181 2.5 mL
22-010-1175 0.05 mL
22-010-1176 0.1 mL
22-010-1178 0.5 mL
22-010-1183 10 mL
22-010-1184 12.5 mL
22-010-1185 25 mL
22-010-1186 50 mL
22-010-1177 0.2 mL
22-010-1179 1.0 mL
22-010-1180 1.25 mL
22-010-1182 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 22-010-1181 Supplier Globe Scientific Supplier No. 3926S
Add to Cart
Add to Cart

For use with Diamond Rv-Pette PRO™ Repeat Volume Pipettor.

  • Standard color coding ensures easy volume identification
  • Safe, secure choice for handling a wide range of liquids including those that are viscous, infectious, and volatile
  • Tested during the automated manufacturing process
  • Available in 12 sizes for dispensing volumes from 1μL to 5500μL
  • Available non-sterile and sterile
  • Sterile tips are individually wrapped and certified to be RNase, DNase, ATP, and pyrogen free for most critical applications

Specifications

Volume (Metric) 2.5 mL
Filtered Non-filtered
Sterility Sterile
Color Natural
Format Individually Wrapped
Disposable Single-use
For Use With (Equipment) Universal Fit Repeater Pipettor
Graduated No
Product Line Diamond RV-Pette PRO
Material Polypropylene, Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.