Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamyl Phthalate 98.0+%, TCI America™
SDP

Catalog No. P0291100G Shop All Tokyo Chemical Industry (TCI) Products
missing translation for 'changeView'
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0291100G 100 g
P029125G 25 g
P0291500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. P0291100G Supplier TCI America missing translation for 'supplierNo' P0291100G
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Chemical Identifiers

CAS 131-18-0
Molecular Formula C18H26O4
Molecular Weight (g/mol) 306.402
MDL Number MFCD00041934
InChI Key IPKKHRVROFYTEK-UHFFFAOYSA-N
Synonym dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester
PubChem CID 8561
ChEBI CHEBI:34680
IUPAC Name dipentyl benzene-1,2-dicarboxylate
SMILES CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC

Specifications

Color Yellow
Boiling Point 342°C
Quantity 100 g
UN Number 3082
Formula Weight 306.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamyl Phthalate
RTECSNumber TI1930000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.