Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIAPH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552173 Supplier Invitrogen™ Supplier No. PA552173
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGSEEPGGGR SNKRSAGNRA ANEEETKNKP KLNIQIKTLA DDVRDRITSF RKSTVKKEKP LIQHPIDSQV AMSEFPAAQP LYDERSLNLS EKEVLDLFEK MMEDMNLNEE KKAPLRNKDF TTKREMVVQY ISAT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 39%.

The product of this gene belongs to the diaphanous subfamily of the formin homology family of proteins. This gene may play a role in the development and normal function of the ovaries. Defects in this gene have been linked to premature ovarian failure 2. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIAPH2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DIAPH2
Gene Accession No. O60879, O70566
Gene Alias DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1
Gene Symbols DIAPH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DIAPH2. Recombinant protein control fragment (Product #RP-92145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1730, 317189, 54004
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.