Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatomaceous Earth, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. MK3371212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-3371-212 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-3371-212 Supplier Avantor Supplier No. 337105

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

    Suitable for use in Pesticide Residue Analysis after Extraction with Petroleum Ether

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 2.5 kg
Particle Size 48 Micron
Specific Gravity 1L = 2.30kg
Packaging Poly Pail
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Diatomaceous Earth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.