Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatomaceous Earth, Non-washed, ∽90% SiO2, MP Biomedicals™

Catalog No. ICN15760691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15760691 1 kg
ICN15760694 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15760691 Supplier MP Biomedicals, Inc Supplier No. 0215760691
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Diatomaceous silica
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Powder
Color Gray to White
Quantity 1 kg
Assay Percent Range ∽90%
Formula Weight 60.04
Chemical Name or Material Diatomaceous Earth, Non-washed
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.