Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazoxide 98.0+%, TCI America™
SDP

Catalog No. D54021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54021G 1 g
D5402250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54021G Supplier TCI America Supplier No. D54021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 364-98-7
Molecular Formula C8H7ClN2O2S
Molecular Weight (g/mol) 230.67
MDL Number MFCD00078578
InChI Key GDLBFKVLRPITMI-UHFFFAOYSA-N
Synonym diazoxide, hyperstat, proglycem, hypertonalum, eudemine, proglicem, dizoxide, mutabase, diazossido, 7-chloro-3-methyl-2h-1,2,4-benzothiadiazine 1,1-dioxide
PubChem CID 3019
ChEBI CHEBI:4495
IUPAC Name 7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
SMILES CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1

Specifications

Melting Point 332°C
Color White-Yellow
Quantity 1 g
Formula Weight 230.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diazoxide
RTECSNumber DK8185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.