Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibasic Potassium Phosphate, Anhydrous, EP, BP, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602144 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-144 500 g Poly Bottle
18-602-145 2.5 kg Poly Bottle
18-602-146 12 kg Poly Pail
18-602-147 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-144 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO204500GM
Add to Cart
Add to Cart

PO204, 7758-11-4, K2HPO4

Spectrum™ Chemical Dibasic Potassium Phosphate, Anhydrous, USP, EP, BP is used as a mineral supplement and is categorized in the United States by the Food and Drug Administration as generally recognized as safe (GRAS). Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

pH 8.5-9.6
CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade Multi-Compendial
Loss on Drying 1%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.