Learn More
Medchemexpress LLC Dibenz[b,f]azocine-5(6H)-pentanoic acid, 11,12-didehydro-δ-oxo- | 1207355-31-4 | MFCD30748340 | 99.7% | 319.36 g/mol | C20H17NO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1209031G
Encompass_Preferred
DBCO-C3-Acid is a strained cyclooctyne-containing click chemistry intermediate for copper-free bioconjugation. It reacts with azide-functionalized molecules via strain-promoted alkyne-azide cycloaddition (SPAAC), enabling construction of antibody-drug conjugate linkers and other bioconjugates; the material is supplied with high reported purity and multiple pack sizes for synthetic scalability.
  • Enables copper-free SPAAC reactions with azide groups.
  • Contains a DBCO moiety for rapid strain-promoted cycloaddition.
  • High purity (approx. 99.7%) suitable for synthetic applications.
  • Supplied in multiple pack sizes including 1 G for scalability.
  • Reported molecular weight 319.36 g/mol and formula C20H17NO3 for accurate stoichiometry.
Catalog No. 50-002-02194