Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo-21-crown 7-Ether 96.0+%, TCI America™
SDP

Catalog No. D28781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28781G Supplier TCI America Supplier No. D28781G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14098-41-0
Molecular Formula C22H28O7
Molecular Weight (g/mol) 404.459
MDL Number MFCD00068646
InChI Key JKCQOMAQPUYHPL-UHFFFAOYSA-N
Synonym dibenzo-21-crown-7, dibenzo-21-crown 7-ether, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, dibenzo-21-crown7-ether, dibenzo-21-crown, 2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro
PubChem CID 585932
SMILES C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

Specifications

Melting Point 107°C
Quantity 1 g
Formula Weight 404.46
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo-21-crown 7-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.