Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo-24-crown 8-Ether 98.0+%, TCI America™
SDP

Catalog No. D18305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18305G 5 g
D18301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18305G Supplier TCI America Supplier No. D18305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14174-09-5
Molecular Formula C24H32O8
Molecular Weight (g/mol) 448.51
MDL Number MFCD00005101
InChI Key UNTITLLXXOKDTB-UHFFFAOYSA-N
PubChem CID 84238
IUPAC Name 2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹⁷]dotriaconta-1(32),12,14,16,28,30-hexaene
SMILES C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 448.51
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo-24-crown 8-Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.