Learn More
Sigma Organic Chemistry Dibenzo b dthiophene-4- | 1G | 23985-81-1 | MFCD00961967
Supplier: Sigma Organic Chemistry CBR005111G

Dibenzo b dthiophene-4- , 1G
About this Item:
CAS #: 23985-81-1
MDL #: MFCD00961967
Molecular Weight: 212.27
UNSPSC Code: 12352200
Chemical Formula: C13H8OS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.