Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzofuran-4-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D47711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47711G 1 g
D47715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47711G Supplier TCI America Supplier No. D47711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96706-46-6
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.205
MDL Number MFCD03306027
InChI Key GQYTWBPRZFRASB-UHFFFAOYSA-N
PubChem CID 126043
IUPAC Name dibenzofuran-4-carbaldehyde
SMILES C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C=O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran-4-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.