Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzofuran Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USEPA1100
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1100 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1100 Supplier Agilent Technologies Supplier No. EPA11001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-64-9 , 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 0.63, 99.36
Analyte or Component Names methanol,p-cresol (4-methylphenol)
Element Methanol, p-cresol (4-methylphenol)
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Dibenzofuran Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.