Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dibenzoyl-D-tartaric Acid 98.0+%, TCI America™
SDP

Catalog No. D3826250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3826250G 250 g
D382625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3826250G Supplier TCI America Supplier No. D3826250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
IUPAC Name (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C
Color White
Quantity 250 g
Formula Weight 358.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric Acid
EINECSNumber (3)-1290
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.