Learn More
Dibenzyl diselenide, 95%
Description
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 91°C to 93°C |
| Quantity | 5 g |
| UN Number | UN3283 |
| Beilstein | 1877745 |
| Solubility Information | Soluble in water. |
| Formula Weight | 340.18 |
| Percent Purity | 95% |
| Chemical Name or Material | Dibenzyl diselenide |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.