Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl Disulfide 98.0+%, TCI America™
SDP

Catalog No. B0417500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0417500G 500 g
B041725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0417500G Supplier TCI America Supplier No. B0417500G

Chemical Identifiers

CAS 150-60-7
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.386
MDL Number MFCD00004783
InChI Key GVPWHKZIJBODOX-UHFFFAOYSA-N
Synonym dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
PubChem CID 9012
ChEBI CHEBI:72752
IUPAC Name (benzyldisulfanyl)methylbenzene
SMILES C1=CC=C(C=C1)CSSCC2=CC=CC=C2

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 500 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Disulfide
EINECSNumber (3)-1136
RTECSNumber JO1750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.