Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl L-Glutamate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D483925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D483925G 25 g
D48395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D483925G Supplier TCI America Supplier No. D483925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4561-10-8
Molecular Formula C19H22ClNO4
Molecular Weight (g/mol) 363.84
MDL Number MFCD00035103
InChI Key GRGJVECUQLAEDM-LMOVPXPDSA-N
Synonym s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl
PubChem CID 199613
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N.Cl

Specifications

Melting Point 104°C
Color White
Quantity 25 g
Formula Weight 363.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl L-Glutamate Hydrochloride
RTECSNumber MA0885000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.