Learn More
Sigma Aldrich Dibenzyl N,N-diethylphosphoramidite
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Dibenzyl n,n-diethylphosphoramidite is repoted to be suitable reagent for the efficient 1h-tetrazole-catalyzed αphosphite-triesterα phosphorylation of the protected serine derivatives.dibenzyl n,n-diethylphosphoramidite may be used:
- As phosphitylating reagent in the synthesis of glycosyl phosphites and phosphates
- For phosphorylating the alcohols
- In the preparation of 6-dibenzylphosphate-1,3,5-tri-o-pmb-myo-inositol
Specifications
Specifications
| CAS | 67746-43-4 |
| Density | 1.06 g/mL (at 25°C (literature)) |
| Molecular Formula | C18H24NO2P |
| Refractive Index | n20/D 1.545 (literature) |
| Linear Formula | (C2H5)2 NP(OCH2C6H5)2 |
| MDL Number | MFCD00191987 |
| Quantity | 1 g |
| Synonym | TTC |
| Molecular Weight (g/mol) | 317.36 |
| Percent Purity | 85% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.