Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl Phosphate 98.0+%, TCI America™
SDP

Catalog No. P112025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P112025G 25 g
P11205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P112025G Supplier TCI America Supplier No. P112025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1623-08-1
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.244
MDL Number MFCD00004775
InChI Key HDFFVHSMHLDSLO-UHFFFAOYSA-N
Synonym dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
PubChem CID 74189
IUPAC Name dibenzyl hydrogen phosphate
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2

Specifications

Melting Point 81°C
Color White
Quantity 25 g
Formula Weight 278.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Phosphate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.