Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl Sebacate 98.0+%, TCI America™
SDP

Catalog No. S014725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014725G 25 g
S01475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S014725G Supplier TCI America Supplier No. S014725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 140-24-9
Molecular Formula C24H30O4
Molecular Weight (g/mol) 382.5
MDL Number MFCD00039640
InChI Key DVLXEVMGHXWBOZ-UHFFFAOYSA-N
Synonym Decanedioic Acid Dibenzyl Ester, Sebacic Acid Dibenzyl Ester
PubChem CID 8790
IUPAC Name dibenzyl decanedioate
SMILES C1=CC=C(C=C1)COC(=O)CCCCCCCCC(=O)OCC2=CC=CC=C2

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 382.50
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Dibenzyl Sebacate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.