Learn More
Sigma Organic Chemistry Dibenzyl succinate | 1G | 103-43-5 | MFCD00022034
Supplier: Sigma Organic Chemistry TMT002021G

Dibenzyl succinate , 1G
About this item:
CAS #: 103-43-5
MDL #: MFCD00022034
Chemical Formula: C18H18O4
Molecular Weight: 298.33
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.