Learn More
Sigma Organic Chemistry Dibenzylamine | 100G | 103-49-1 | MFCD00004770
Supplier: Sigma Organic Chemistry D34108100G
Dibenzylamine, 100G
About This Item:
Linear Formula: (C6H5CH2)2NH
Molecular Weight: 197.28
CAS Number: 103-49-1
MDL Number: MFCD00004770
UNSPSC Code: 12352100
Purity: 0.97
Boiling Point: 300 C (lit.)
Melting Point: -26 C (lit.)
Density: 1.026 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.574 (lit.)
Storage: room temp
EINECS Number: 203-117-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.