Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl ether, puriss p.a., ≥99.5% (GC), Solstice

Catalog No. 60048419 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-419 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-419 Supplier Solstice Supplier No. 344621L

missing translation for 'validMainframeMsg'

Puriss. p.a., low in aromatic compounds,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-96-1
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00009461
InChI Key DURPTKYDGMDSBL-UHFFFAOYSA-N
Synonym di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
PubChem CID 8909
IUPAC Name 1-butoxybutane
SMILES CCCCOCCCC

Specifications

Density 0.764g/mL at 25°C
Flash Point 25°C (77°F)
Ignition Point 365°F
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2O
Quantity 1 L
UN Number UN1149
Beilstein 1732752
Vapor Density 4.48 (vs air)
Vapor Pressure 4.8mmHg (20°C)
Formula Weight 130.23g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Dibutyl ether
Show More Show Less
Hazard Statement P210-P273-P280-P304 + P340 + P312-P337 + P313-P403 + P235
DOTInformation class 3 - PG 3 - Dibutyl ethers
RTECSNumber 205-575-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.