Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl phthalate

Catalog No. 502898566
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-289-8566 4 L
50-290-3191 18 L
50-183-4308 100 mL
50-183-4311 500 mL
NC1014483 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-289-8566 Supplier Sigma Aldrich Supplier No. 5249804L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyl phthalate
TRUSTED_SUSTAINABILITY

Specifications

CAS 84-74-2
Melting Point -35°C (lit.)
Density 1.043 g/mL (at 25°C (literature))
Boiling Point 340°C (lit.)
Molecular Formula C16H22O4
Refractive Index n20/D 1.492 (literature)
Linear Formula C6H4-1,2-[CO2(CH2)3 CH3]2
MDL Number MFCD00009441
Quantity 4 L
Synonym n-Butyl phthalate; DBP; Phthalic acid dibutyl ester
Molecular Weight (g/mol) 278.34
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.