Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl sebacate, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012441 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2441 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2441 Supplier MilliporeSigma Supelco Supplier No. 808651ML
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-43-3
Density 0.936 g/mL (at 25°C)
Flash Point 178°C
Physical Form Neat
Linear Formula [-(CH2)4 CO2(CH2)3 CH3]2
MDL Number MFCD00027218
Boiling Point 178°C to 179°C (3 mmHg)
Synonym Decanedioic acid dibutyl ester; Sebacic acid dibutyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 314.46
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.