Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutylamine, Approx. 99%, Spectrum™ Chemical
SDP

Catalog No. 18610177 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-177 200 L Amber Glass Bottle
18-610-174 100 mL Amber Glass Bottle
18-610-178 500 mL Amber Glass Bottle
18-610-173 1 L Amber Glass Bottle
18-610-175 2.5 L Amber Glass Bottle
18-610-176 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-610-177 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1090200LTBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-92-2

Specifications

CAS Max % 1
Boiling Point 159°C to 160°C
Packaging Amber Glass Bottle
Quantity 200 L
Molecular Weight (g/mol) 129.25
Percent Purity ∼99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dibutylamine

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.