Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin Dilaurate 95.0+%, TCI America™
SDP

Catalog No. D0303100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0303100G 100 g
D030325G 25 g
D0303500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0303100G Supplier TCI America Supplier No. D0303100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77-58-7
Molecular Formula C32H64O4Sn
MDL Number MFCD00008963

Specifications

Color Yellow
Physical Form Liquid
Quantity 100 g
UN Number 2788
Formula Weight 631.57
Percent Purity ≥95.0% (W)
Chemical Name or Material Dibutyltin Dilaurate
EINECSNumber (2)-2330
RTECSNumber WH7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.