Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin Oxide 95.0+%, TCI America™
SDP

Catalog No. D0305100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0305100G 100 g
D030525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0305100G Supplier TCI America Supplier No. D0305100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Color White
Quantity 100 g
UN Number 3146
Formula Weight 248.94
Percent Purity ≥95.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyltin Oxide
EINECSNumber (2)-2031
RTECSNumber WH7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.