Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicamba Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USHB1601
Encompass
Change view
Click to view available options
Quantity:
4 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HB-160-1 4 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HB-160-1 Supplier Agilent Technologies Supplier No. HB1601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1918-00-9 , 67-56-1
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
InChI Key IWEDIXLBFLAXBO-UHFFFAOYSA-N
PubChem CID 3030
ChEBI CHEBI:81856
IUPAC Name 3,6-dichloro-2-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1C(=O)O)Cl)Cl

Specifications

CAS Max % 99.98, 0.01
Analyte or Component Names Methanol,dicamba
Element Methanol,dicamba
Solvent or Matrix Methanol
Quantity 4 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Dicamba Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.