Learn More
Medchemexpress LLC HY-135737 500mg Medchemexpress, Dicaprylyl carbonate CAS:1680-31-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135737500MG
Encompass_Preferred
Medchemexpress, HY-135737 500mg Dicaprylyl carbonate CAS:1680-31-5 Dicaprylyl carbonate, a solid, plant-derived fat, is a dry emollient. Dicaprylyl carbonate has excellent dermatological compatibility and a comprehensive performance profile, such as solubilizing and dispersing ability for sun-care filters. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0351
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.