Learn More
Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%
Description
(Acetylacetonato)dicarbonylrhodium(I) is used in in situ formation of a floors-soluble hydroformylation catalyst of interest in molecular engineering. It is also used as a catalyst for various carbonylation reactions, silylcarbocyclizations, conjugate additions to eons, carbamoylstannation, and reduction of aromatic nitro compounds. It is also used in Hydroformylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 144°C to 147° (sublimation) |
| Color | Green |
| Odor | Odorless |
| Assay Percent Range | 99% |
| Quantity | 5 g |
| Solubility Information | Soluble in acetone |
| Formula Weight | 258.04 |
| Physical Form | Crystalline |
| Chemical Name or Material | Dicarbonyl(2, 4-pentanedionato)rhodium(I) |
Safety and Handling
| Hazard Category | H228-H301-H317-H319 |
| Precautionary Statement | P210-P240-P241-P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P330-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 6.1; Packaging Group: III |
| EINECSNumber | 238-947-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.