Learn More
Invitrogen™ Dicer Polyclonal Antibody

Description
Dicer Polyclonal Antibody for IHC (P), Flow, ELISA
Specifications
Specifications
| Antigen | Dicer |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 50% glycerol and 0.025% sodium azide |
| Gene | DICER1 |
| Gene Accession No. | Q9UPY3 |
| Gene Alias | 1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; Dicer; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; Dicer1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; Helicase MOI; helicase with RNAse motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2 |
| Gene Symbols | DICER1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.