Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(1,10-phenanthroline)copper(II) 98.0+%, TCI America™
SDP

Catalog No. D38911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38911G 1 g
D38915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38911G Supplier TCI America Supplier No. D38911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14783-09-6
Molecular Formula C12H8Cl2CuN2
Molecular Weight (g/mol) 314.656
MDL Number MFCD00134224
InChI Key PHECBGBJCLDCMF-UHFFFAOYSA-L
Synonym (1,10-Phenanthroline)copper(II) Dichloride
PubChem CID 6100472
IUPAC Name dichlorocopper;1,10-phenanthroline
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl[Cu]Cl

Specifications

Color Green-Yellow
Quantity 1 g
Formula Weight 314.66
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloro(1,10-phenanthroline)copper(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.