Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro(dimethylamino)phosphine, 98%

Catalog No. AAH2785614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2785614 25 g
AAH2785606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2785614 Supplier Thermo Scientific Chemicals Supplier No. H2785614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 683-85-2 | C2H6Cl2NP | 145.951 g/mol

Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-85-2
Molecular Formula C2H6Cl2NP
Molecular Weight (g/mol) 145.951
MDL Number MFCD00013613
InChI Key XPWWDZRSNFSLRQ-UHFFFAOYSA-N
Synonym dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
PubChem CID 136483
IUPAC Name N-dichlorophosphanyl-N-methylmethanamine
SMILES CN(C)P(Cl)Cl

Specifications

Density 1.271
Boiling Point 150°C
Flash Point −23°C (−9°F)
Linear Formula (CH3)2NPCl2
Quantity 25 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 145.96
Percent Purity 98%
Chemical Name or Material Dichloro(dimethylamino)phosphine
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.