Learn More
Sigma Organic Chemistry Dichloroacetic acid | 100ML | 79-43-6 | MFCD00004223
Supplier: Sigma Organic Chemistry D54702100ML

Dichloroacetic acid, 100ML
About This Item:
Linear Formula: Cl2CHCOOH
Molecular Weight: 128.94
CAS Number: 79-43-6
MDL Number: MFCD00004223
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 194 C (lit.)
Melting Point: 9-11 C (lit.)
Density: 1.563 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.466 (lit.)
Storage: 2-8C
EINECS Number: 201-207-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.