Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.5% min., ACS, MilliporeSigma™

Catalog No. MDX08355 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MDX08355 20 L
MDX08356 500 mL
MDX08353 4 L
MDX083520 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MDX08355 Supplier MilliporeSigma Supplier No. DX08355
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Quantity 20 L
Physical Form Liquid
Assay Percent Range 99.5% min.
Formula Weight 84.93
Chemical Name or Material Dichloromethane, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.